ENA181911844 Enamine

3-(3-fluorophenyl)-2-oxopropanoic acid

  • MDL number MFCD09729862
  • Cas Number : 95041-89-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37578

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37578
Availability details for
Quantity
  • Availability
  • Add to Cart