ENA181911852 Enamine

N-(4-ethylbenzyl)cyclopropanamine

  • MDL number MFCD07409061
  • Cas Number : 892576-76-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33139

Safety Information

msds https://www.enaminestore.com/catalog/EN300-33139
Availability details for
Quantity
  • Availability
  • Add to Cart