ENA181911857 Enamine

N-(1,1'-biphenyl-4-ylmethyl)-N-methylamine

  • MDL number MFCD07410264
  • Cas Number : 110931-72-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33199

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-33199
pictograms GHS05,GHS07,GHS09
Availability details for
Quantity
  • Availability
  • Add to Cart