ENA181911861 Enamine

4-azaspiro[bicyclo[2.2.2]octane-2,4'-imidazolidine]-2',5'-dione

  • MDL number MFCD00047561
  • Cas Number : 37874-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-38877

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-38877
Availability details for
Quantity
  • Availability
  • Add to Cart