ENA181911867 Enamine

2-fluoro-4-(thiophen-2-yl)benzoic acid

  • MDL number MFCD09042180
  • Cas Number : 505082-90-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-38883

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-38883
Availability details for
Quantity
  • Availability
  • Add to Cart