ENA181911875 Enamine

5-(2-hydroxyethyl)-2-oxo-6-(trifluoromethyl)-1,2-dihydropyridine-3-carboxamide

  • MDL number MFCD09731037
  • Cas Number : 953882-05-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-38908

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-38908
Availability details for
Quantity
  • Availability
  • Add to Cart