ENA181911878 Enamine

4-{[4-(3-methylphenyl)piperazin-1-yl]methyl}aniline

  • MDL number MFCD09816235
  • Cas Number : 1016711-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-38913

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-38913
Availability details for
Quantity
  • Availability
  • Add to Cart