ENA181911888 Enamine

3-chloro-N-[4-(pyrrolidine-1-sulfonyl)phenyl]propanamide

  • MDL number MFCD08079208
  • Cas Number : 895691-70-0
  • Molecular Weight : 316.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-25825

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25825
Availability details for
Quantity
  • Availability
  • Add to Cart