ENA181911894 Enamine

2-amino-5-bromophenol

  • MDL number MFCD03095028
  • Cas Number : 38191-34-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138...140
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-38932

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-38932
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart