ENA181911908 Enamine

4-bromo-3-sulfamoylbenzoic acid

  • MDL number MFCD00625736
  • Cas Number : 59815-19-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-38952

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-38952
Availability details for
Quantity
  • Availability
  • Add to Cart