ENA181911932 Enamine

3-chloro-5-(2-chlorophenyl)-4-ethyl-4H-1,2,4-triazole

  • MDL number MFCD11099603
  • Cas Number : 1083381-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-38990

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-38990
Availability details for
Quantity
  • Availability
  • Add to Cart