ENA181911936 Enamine

N-(4-acetylphenyl)-N'-cyclopropylurea

  • MDL number MFCD06117650
  • Cas Number : 847477-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 161...163
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36467

Safety Information

msds https://www.enaminestore.com/catalog/EN300-36467
Availability details for
Quantity
  • Availability
  • Add to Cart