ENA181911937 Enamine

3-[5-(2-furyl)-1,3-oxazol-2-yl]propanoic acid

  • MDL number MFCD09804817
  • Cas Number : 1016674-88-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140...142
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-38997

Safety Information

msds https://www.enaminestore.com/catalog/EN300-38997
Availability details for
Quantity
  • Availability
  • Add to Cart