ENA181911938 Enamine

5,7-dimethyl-[1,2,4]triazolo[4,3-c]pyrimidine-3-thiol

  • MDL number MFCD09880773
  • Cas Number : 74537-87-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36474

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36474
Availability details for
Quantity
  • Availability
  • Add to Cart