ENA181911940 Enamine

ethyl 2-[(2-methylpropyl)amino]acetate

  • MDL number MFCD11099609
  • Cas Number : 3182-87-4
  • Molecular Weight : 159.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-39004

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39004
Availability details for
Quantity
  • Availability
  • Add to Cart