ENA181911941 Enamine

2-[(methoxycarbonyl)(methyl)amino]acetic acid

  • MDL number MFCD11099610
  • Cas Number : 116714-27-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-39005

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39005
Availability details for
Quantity
  • Availability
  • Add to Cart