ENA181911942 Enamine

2-(ethylamino)-N,N-dimethylacetamide

  • MDL number MFCD09811850
  • Cas Number : 97454-95-0
  • Molecular Weight : 130.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-39006

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39006
Availability details for
Quantity
  • Availability
  • Add to Cart