ENA181911943 Enamine

2-(ethylamino)-N-methylacetamide

  • MDL number MFCD09808153
  • Cas Number : 225229-03-8
  • Molecular Weight : 116.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-39007

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39007
Availability details for
Quantity
  • Availability
  • Add to Cart