ENA181911945 Enamine

1-(5-aminopyridin-2-yl)piperidine-3-carboxamide

  • MDL number MFCD09736214
  • Cas Number : 953726-10-8
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39009

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39009
Availability details for
Quantity
  • Availability
  • Add to Cart