ENA181911952 Enamine

{[5-(trifluoromethyl)pyridin-2-yl]amino}acetic acid

  • MDL number MFCD08444829
  • Cas Number : 923216-18-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134...136
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39030

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39030
Availability details for
Quantity
  • Availability
  • Add to Cart