ENA181911953 Enamine

2-(ethylamino)-N-(4-methoxyphenyl)acetamide

  • MDL number MFCD05263152
  • Cas Number : 743444-49-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39031

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39031
Availability details for
Quantity
  • Availability
  • Add to Cart