ENA181911955 Enamine

3-(isonicotinoylamino)propanoic acid

  • MDL number MFCD00463120
  • Cas Number : 147218-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 239...241
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39033

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39033
Availability details for
Quantity
  • Availability
  • Add to Cart