ENA181911959 Enamine

[3-(dimethylamino)propyl](2-methoxyethyl)amine

  • MDL number MFCD11099617
  • Cas Number : 84176-68-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39037

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39037
Availability details for
Quantity
  • Availability
  • Add to Cart