ENA181911968 Enamine

1-[4-(2-methoxyethoxy)phenyl]methanamine

  • MDL number MFCD09816053
  • Cas Number : 102196-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39047

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-39047
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart