ENA181911978 Enamine

N,N-diethyl-N'-isobutylethane-1,2-diamine

  • MDL number MFCD11099623
  • Cas Number : 50342-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39063
Availability details for
Quantity
  • Availability
  • Add to Cart