ENA181911982 Enamine

N-(piperidin-3-yl)methanesulfonamide

  • MDL number MFCD09810664
  • Cas Number : 944068-21-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-39067

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39067
Availability details for
Quantity
  • Availability
  • Add to Cart