ENA181911983 Enamine

3-(3,5-dimethyl-1H-pyrazol-1-yl)aniline

  • MDL number MFCD09696862
  • Cas Number : 1007303-77-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-39068

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39068
Availability details for
Quantity
  • Availability
  • Add to Cart