ENA181911984 Enamine

1-(2-nitrophenyl)-3-(propan-2-yl)urea

  • MDL number MFCD03208832
  • Cas Number : 347907-99-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39070

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39070
Availability details for
Quantity
  • Availability
  • Add to Cart