ENA181911989 Enamine

1-(3-cyanophenyl)-3-(propan-2-yl)urea

  • MDL number MFCD11099418
  • Cas Number : 1038255-83-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36725

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36725
Availability details for
Quantity
  • Availability
  • Add to Cart