ENA181911992 Enamine

2-[(2,2,2-trifluoroethyl)amino]ethan-1-ol

  • MDL number MFCD10697861
  • Cas Number : 371-99-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39081

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39081
Availability details for
Quantity
  • Availability
  • Add to Cart