ENA181911997 Enamine

4-(2-amino-1,3-thiazol-4-yl)-N,N-dimethylbenzenesulfonamide

  • MDL number MFCD03444373
  • Cas Number : 610275-82-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 243...245
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39088

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39088
Availability details for
Quantity
  • Availability
  • Add to Cart