ENA181912002 Enamine

1-(5-methyl-2-phenyl-1H-imidazol-4-yl)ethan-1-one

  • MDL number MFCD01098518
  • Cas Number : 28824-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-39095

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39095
Availability details for
Quantity
  • Availability
  • Add to Cart