ENA181912004 Enamine

N-(2-aminoethyl)-1-phenylmethanesulfonamide

  • MDL number MFCD04364577
  • Cas Number : 190257-50-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-39098

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39098
Availability details for
Quantity
  • Availability
  • Add to Cart