ENA181912005 Enamine

[2-amino-1-(thiophen-3-yl)ethyl]diethylamine

  • MDL number MFCD09731793
  • Cas Number : 953892-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-39099

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39099
Availability details for
Quantity
  • Availability
  • Add to Cart