ENA181912012 Enamine

3-chloro-1-methyl-1H-indole-2-carboxylic acid

  • MDL number MFCD11099635
  • Cas Number : 1043420-84-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 233...235
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39111

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39111
Availability details for
Quantity
  • Availability
  • Add to Cart