ENA181912027 Enamine

N-[2-(pyridin-2-yl)ethyl]piperidine-4-carboxamide dihydrochloride

  • MDL number MFCD11099420
  • Cas Number : 1170009-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Product Link https://www.enaminestore.com/catalog/EN300-36758

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36758
Availability details for
Quantity
  • Availability
  • Add to Cart