ENA181912029 Enamine

3-methylthieno[2,3-b]pyridine-2,5-dicarboxylic acid

  • MDL number MFCD10586460
  • Cas Number : 949984-37-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36761

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36761
Availability details for
Quantity
  • Availability
  • Add to Cart