ENA181912031 Enamine

3-phenylmethanesulfonylpropan-1-amine

  • MDL number MFCD09942626
  • Cas Number : 933699-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-39147

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39147
Availability details for
Quantity
  • Availability
  • Add to Cart