ENA181912035 Enamine

4-[4-(3-methylphenyl)piperazin-1-yl]butan-1-amine

  • MDL number MFCD09723567
  • Cas Number : 90931-05-8
  • Molecular Weight : 247.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-36764

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36764
Availability details for
Quantity
  • Availability
  • Add to Cart