ENA181912037 Enamine

2-(4-aminophenyl)-2-phenylacetonitrile hydrochloride

  • MDL number MFCD00065214
  • Cas Number : 69833-17-6
  • Molecular Weight : 244.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-39156

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39156
Availability details for
Quantity
  • Availability
  • Add to Cart