ENA181912038 Enamine

2-(4-aminopiperidin-1-yl)-N-propylacetamide

  • MDL number MFCD09802455
  • Cas Number : 1016510-04-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39157

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39157
Availability details for
Quantity
  • Availability
  • Add to Cart