ENA181912044 Enamine

1-(4-isobutoxy-3-methoxyphenyl)methanamine

  • MDL number MFCD09729547
  • Cas Number : 854185-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88...90
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36765

Safety Information

msds https://www.enaminestore.com/catalog/EN300-36765
Availability details for
Quantity
  • Availability
  • Add to Cart