ENA181912054 Enamine

1-isonicotinoylpiperidin-4-one oxime

  • MDL number MFCD09951206
  • Cas Number : 1017043-97-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 186...188
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39175

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39175
Availability details for
Quantity
  • Availability
  • Add to Cart