ENA181912060 Enamine

2-bromo-N-(2-chlorophenyl)-3-methylbutanamide

  • MDL number MFCD04406119
  • Cas Number : 391220-24-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114...116
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36783

Safety Information

msds https://www.enaminestore.com/catalog/EN300-36783
Availability details for
Quantity
  • Availability
  • Add to Cart