ENA181912061 Enamine

4-(2-bromoacetyl)benzene-1-sulfonamide

  • MDL number MFCD08689427
  • Cas Number : 944-33-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39186

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39186
Availability details for
Quantity
  • Availability
  • Add to Cart