ENA181912064 Enamine

1-benzylpiperidin-3-amine

  • MDL number MFCD04038444
  • Cas Number : 60407-35-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-36785
Availability details for
Quantity
  • Availability
  • Add to Cart