ENA181912082 Enamine

tert-butyl N-(1-benzylpiperidin-3-yl)carbamate

  • MDL number MFCD09261310
  • Cas Number : 478828-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39215

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39215
Availability details for
Quantity
  • Availability
  • Add to Cart