ENA181912084 Enamine

tert-butyl N-(3-aminopropyl)-N-[(pyridin-3-yl)methyl]carbamate

  • MDL number MFCD11099660
  • Cas Number : 1019452-24-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39229

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39229
Availability details for
Quantity
  • Availability
  • Add to Cart