ENA181912090 Enamine

[1-(thiomorpholin-4-yl)cyclopentyl]methanamine

  • MDL number MFCD11099664
  • Cas Number : 1042651-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39247

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39247
Availability details for
Quantity
  • Availability
  • Add to Cart