ENA181912115 Enamine

2-methyl-4-nitrobenzene-1-sulfonyl chloride

  • MDL number MFCD09050619
  • Cas Number : 21320-90-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68-70
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36815

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36815
Availability details for
Quantity
  • Availability
  • Add to Cart